VWR® VisiScope® TL384B FL1 B, Fluorescence Microscope
Leverandør: VWR Collection
630-3242EA 30850 NOK
630-3242
VWR® VisiScope® TL384B FL1 B, Fluorescence Microscope
Mikroskop
These fluorescence microscopes are designed for routine lab applications.
  • Observation methods: Brightfield and LED fluorescence
  • Energy saving LED technology with higher light intensity for bright images and improved colour fidelity
  • Automatic light control and adjustment
  • Rackless stage with belt-driven mechanism allows a smooth movement
  • Five objectives with anti-fungus treatment
This extremely powerful LED fluorescence illuminator is combined with blue excitation filter set for the visualisation of the following fluorochromes: Acridine Yellow, AcridineMore Product Information
Bestill nå
SPESIFIKASJONER
Modell TL384B FL1 B
Environmentally Preferable
Kontrastteknikk Lysfelt og fluorescens
Head Dreibar 360°, vinklet 30°
Eyepieces WF 10×/20 mm
Nosepiece 5 posisjoner
Objectives 4×, 10×, 20×, 40×, 60×, 100× (i)
Stage 233×147 mm (movement: 78×54 mm)
Condenser Abbe N.A. 1.25, with iris diaphragm, focusable and centerable
Fokuseringssystem Koaksial grov- og finfokusering
Illumination Transmitted illumination (fixed Koehler type): LED (white 3,6 W); Epi-illumination: High power blue LED with brightness control; 1-position filter holder (blue filter included)
B×D×H 210×370×480 mm
Vekt 5 kg