VWR® VisiScope® IT600 FLD, Fluorescence Microscope, Inverted Trinocular
Leverandør: VWR Collection
630-3267EA 0 NOK
630-3267
VWR® VisiScope® IT600 FLD, Fluorescence Microscope, Inverted Trinocular
Mikroskop
Routine inverted fluorescence microscopes for transmitted brightfield, darkfield, phase contrast and fluorescence observations. Ideal for tissue culture research in clinical and biotechnology laboratories.
  • No warm-up or cool-down times for 60-minute time saving
  • High-efficiency LED illumination with brightness control
  • Interchangeable LED filter sets
  • Condenser can be removed/rotated to extend the working distance up to 220 mm
  • Full range of objectives to choose from for highest flexibility
These extremely powerful LED fluorescence illuminators areMore Product Information