VWR® VisiScope® BL224PL T1, Digital Microscope
Leverandør: VWR Collection
630-3290EA 20400 NOK
630-3290
VWR® VisiScope® BL224PL T1, Digital Microscope
Mikroskop
Educational and laboratory microscope with integrated camera and removable tablet.
  • Die-cast frame for high stability and ergonomics during transmitted light observations
  • Tablet on the microscope rotatable in all directions and easily detached
  • Large opening behind the revolving nosepiece can be used as a handle
  • N-plan objectives
  • White illumination with light intensity controlled via a knob
  • Efficient LED energy saving lamp
Multilingual tablet with touch screen LCD for wireless or Bluetooth® connection and powered by a two-cell Li-Ion battery. Please seeMore Product Information