Digital microscopes, Emspira 3
Leverandør: LEICA MICROSYSTEMS
10450851_P 10450936_P 10450923
630-3381EA 0 NOK
630-3381 630-3382 LEIM10450923
Digital microscopes, Emspira 3
Mikroskop
Emspira 3 digital microscope empowers users to streamline inspection processes, cover inspection needs flexibly, and work with confidence. An ideal partner for manipulation, quality control, inspection and research applications.
  • Flexibility thanks to the modular design and large variety of accessories
  • Ease of use - create overlays and reference images for comparison
  • Future proof - capture directly to USB stick or network folder or send images via email
  • Intuitive, customisable UI with mouse controls
  • High quality, robust and antimicrobial housing AgTreat™ -