Inverted LED microscopes, DMi1
Leverandør: LEICA MICROSYSTEMS
11526234 11526235 11526236
630-3434EA 59100 NOK
630-3434 630-3435 630-3436
Inverted LED microscopes, DMi1
Mikroskop
Get what you need and increase efficiency in your live cell imaging workflow with the Leica DMi1 inverted microscope.
  • Easy to use
  • Easy phase contrast
  • Cool, colour safe LED illumination
  • HD imaging option available
The unique flexibility of the Leica DMi1 allows you to configure the microscope for your needs with flexible condenser options and digital imaging documentation features; creating a solution that is just right for your lab.
Easy to use operation that requires minimal training and maintenance so you can fully concentrate on your research.More Product Information
Bestill nå
SPESIFIKASJONER
Modell DM i1 package 1 DM i1 package 3 DM i1 package 6
Environmentally Preferable
Okularer 2 exchangeable eyepieces 10×/20 eyepiece with eyeguard and 10×/20 focusing eyepiece with eyeguard 2 exchangeable eyepieces 10×/20 eyepiece with eyeguard and 10×/20 focusing eyepiece with eyeguard 2 exchangeable eyepieces 10×/20 eyepiece with eyeguard and 10×/20 focusing eyepiece with eyeguard
Arbeidsavstand 40/50 mm 80 mm 80 mm
Kondensor S40 [4-position slider for S40 condenser (BF, Ph0, Ph1, Ph2)] S80 (BF, Ph1) [4-position slider for S80 condenser (BF, Ph0, Ph1, Ph2)] S40 [4-position slider for S40 condenser (BF, Ph0, Ph1, Ph2)]
Belysning LED LED LED
Kamera No connection port No connection port FLEXACAM C1 (12 MP) with multiple ports (HDMI, USB, Ethernet)