Biological microscopes, fixed Koehler halogen and LED, BA 210 Elite
Leverandør: MOTIC
1100100403761 1100100403774 1100100403763 1100100403771
630-3442EA 19375 NOK
630-3442 630-3445 630-3443 630-3444
Biological microscopes, fixed Koehler halogen and LED, BA 210 Elite
Mikroskop
Microscope for basic work in biology life science. Fixed Koehler set-up, rackless stage, nice plan achromatic optics. The Motic BA 210 is robust and ready to use with pre-installed set-up of optics and halogen illumination, interchangeable with a LED module (LED to be purchased separately).
  • Binocular or trinocular head, 30° inclined, 360° rotating
  • Eyepieces N-WF 10×/20 mm with dioptre adjustment on both eyepieces, ±5 dioptre
  • Nosepiece reversed quadruple
  • Objectives CCIS® EC-plan achromatic, DIN, 4×, 10×, 40×, (100× oil optional)
  • Rackless mechanical stage 150