Measuring magnifiers
Leverandør: ESCHENBACH OPTIK
115410 11547
630-8270EA 4650 NOK
630-8270 630-8271
Measuring magnifiers
Forstørrelsesglass
These technical magnifiers can be used for every standard application.
  • Achromatic lens made of high quality silicate for a distortion-free image with pinpoint clarity
  • Fine focusing via easy-grip focusing ring
  • Housing made from glass fibre reinforced polyamide
There are different precision scales available for use with the magnifiers.
Bestillingsinformasjon: Supplied with standard crosshair scale (no. 115201), measuring range 20 mm, 1/10 mm graduation.