Magnesium ionophore I, Selectophore™, function tested ionophore, Supelco®

Leverandør: Merck

Synonymer: N,N′-Diheptyl-N,N′-dimethyl-1,4-butanediamide, ETH 1117

M-Clarity™ quality level = MQ100

63082-50MG-F
63082-50MG-FEA 1375 NOK
63082-50MG-F
Magnesium ionophore I, Selectophore™, function tested ionophore, Supelco®
Magnesium ionophore I

Magnesium ionophore I (ETH 1117) has been used to prepare calcium-magnesium selective electrode


Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₀H₄₀N₂O₂
MDL nummer: MFCD00043110
CAS-nummer: 75513-72-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab41c0a572a5cd9f153e654e30fe9d04999dffde&vid=7156165c7d78d92bd0489548cada6f4a174dfffa&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR