Hulromsglider
Leverandør: Epredia
X1XER315L X2XER208B X1XER301B X1XER302W X2XER201B X2XER202W X2XER203B X1XER308B X1XES306W X1XER305B X1XER303B X1XER311B
631-0463EA 2300 NOK
631-0463 631-1371 631-0451 631-0452 631-0465 631-9423 631-0467 631-0457 631-0464 631-0455 631-0453 631-0460
Hulromsglider
Objektglass Cavity Slides
These autoclavable slides are specially designed for immunofluorescence.
  • Adhesive surface and hydrophobic mask
  • Hydrophobic epoxy- or PTFE-coating
  • (*) with rings or wells
Developped for clinical diagnosis applications.
Bestillingsinformasjon: Available with rings or wells (*).