Staining jars acc. to Coplin
Leverandør: MARIENFELD
4200003
631-0700EA 5200 NOK
631-0700
Staining jars acc. to Coplin
Krukker Fargetrau
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm).
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.