VWR® Q Path®, Slide Cabinet
Leverandør: VWR Collection
631-1066EA 1250 NOK
631-1066
VWR® Q Path®, Slide Cabinet
Objektglassbokser
This cabinet, with lid, can be used in cytology and histology applications.
  • Simple design for easy handling
  • Quick handling of microscope slides
  • Able to hold a large number of items
W×D×H: 84×155×310 mm