VWR® Q Path®, Slide organisers
Leverandør: VWR Collection
631-1068EA 12425 NOK
631-1068 631-1069
VWR® Q Path®, Slide organisers
Objektglassholdere
These organisers enable the archiving of a large number of microscope slides.
  • Robust and stackable
W×D×H: 480×480×140 mm
Leveringsinformasjon: Supplied with a locking rack.