VWR® VisiLight® LED200, Cold Light Source
Leverandør: VWR Collection
631-1085EA 11325 NOK
631-1085
VWR® VisiLight® LED200, Cold Light Source
Lys Kaldlyskilder
This high power 14 W LED source provides a pure-white light (6300 K).
  • Brightness control
  • Illumination (at 100 mm): 70 000 lux
  • Dual gooseneck light guide (L = 500 mm)
W×D: 120×200 mm
Weight: 4,1 kg
Leveringsinformasjon: Supplied with a multi-adapter power supply (EU, UK).