VWR® Premium Plus, Slide Boxes
Leverandør: VWR Collection
631-1248EA 385 NOK
631-1248 631-1242 631-1243 631-1244 631-1245 631-1246 631-1247
VWR® Premium Plus, Slide Boxes
Objektglassbokser
These slide boxes feature multiple labelling areas, and an index card can be used as the lid is transparent.
  • Slide rail design ensures a secure fit
  • Central numbering grid for slide organisation and easy viewing
  • Plastic design eliminates potential rusting of metal parts
  • Sliding lock provides secure storage
  • Hinged lid smoothly opens 180°, sits flat on surface preventing misplacement
  • EVA foam lines the bottom of the box