Magnetglass, Polysine®, Menzel Gläser
Leverandør: Epredia
J2800AMNZ
631-1349EA 610 NOK
631-1349
Magnetglass, Polysine®, Menzel Gläser
Objektglass Adhesjons objektglass
These slides, made using a technology that electrostatically and chemically attracts fresh cell preparations, formalin-fixed Bouin's or alcohol-fixed paraffin-embedded tissues.
  • 90° ground edges with frosted ends for labelling
  • Adhesion superior to glue-, protein- or silane-treated slides
  • Thick tissue sections of 2 to 5 µm
Unaffected by enzyme pre-digestion or heating, tissue loss is minimised in prolonged in-situ DNA hybridisation techniques and in immunocytochemical methods.