VWR®, Low Temperature PC Slide Box
Leverandør: VWR Collection
631-1527EA 510 NOK
631-1527
VWR®, Low Temperature PC Slide Box
Objektglassbokser
Durable and stackable slide box for safe storage and transport of slides.
  • Grooved slide slots separate slides and prevent slide contact
  • Temperature resistant foam lining: Protects slides in low temperatures (−80 °C)
  • Easy slide retrieval, identification sheet, lining, and grooved slots are numbered