Staining racks
Leverandør: KARTELL
354
631-9520EA 330 NOK
631-9520
Staining racks
Stativer for rør Fargestativer
PP.
  • For use with 631-9511
Staining rack with horizontal tray, holds 20 standard slides 76×26 mm.
B×D×H: 86×695×21 mm