Cell density meter, CO 8000 Biowave
Leverandør: Biochrom
80-3000-45
633-0162EA 0 NOK
633-0162
Cell density meter, CO 8000 Biowave
Kolorimeter Multiparameter Kolorimeter
The CO 8000 cell density meter is a small, portable and easy to use instrument for measuring the density of E.coli and yeast cells in suspension at 600 nm.
  • Lightweight device for measuring the cell density of cultures
  • Easy to clean with ethanol or sterilise with formaldehyde or ethylene oxide
  • Long lifetime LED source
  • Rechargeable batteries automatically charged when connected to the mains
  • One month use under normal operating conditions when fully charged offering great flexibility and portability
For use in small research labs, the CO 8000 may beMore Product Information