Hitachi F-7100, High fluorescence spectrophotometer
Leverandør: VWR Collection
5J1-0042
634-1065EA 0 NOK
634-1065
Hitachi F-7100, High fluorescence spectrophotometer
Spektrofotometre og fotometre Fluorenscensspektrofotometere
High fluorescence spectrophotometer with enhanced optical technology and improved performance.
  • Three-dimensional spectra of coumarins for environmental applications
  • Signal-to-noise ratio: 360 (P-P), 1,200 (RMS), 20000 (BG)
  • Robust and reliable high sensitivity detection system
Suitable for use for in research, quality control and education, as well as industrial materials such as LEDs and solar-cell materials, environmental science, food inspection, and life sciences.