VWR®, Macro Cuvettes
Leverandør: VWR Collection
634-1067EA 365 NOK
634-1067
VWR®, Macro Cuvettes
Kyvetter Spektrofotometerkyvetter
Disposable macro cuvettes, four clear sides.
  • Clear and clean optical path
  • Four windows for precise measurements
  • Manufactured under controlled room conditions
Specially designed for fluorescence spectroscopy.