VWR® M4, UV/Visible Spectrophotometer
Leverandør: VWR Collection
634-1227EA 74100 NOK
634-1227
VWR® M4, UV/Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
The M4 single split-beam spectrophotometer, with flashing xenon lamp, is an easy to use, highly reliable and accurate product.
  • Single split-beam design for sample and blank variability that can be monitored at the same time
  • Xenon flash lamp with a very strong energy in the UV and visible range
  • Connects to a computer based on Windows® through various interfaces
Measure absorbance and transmission at any defined wavelength, generates a graph and searches for peaks and valleys, also analyses the graph via built-in methods.
The application can beMore Product Information