VWR® PV4 Visible Spectrophotometer
Leverandør: VWR Collection
634-1088EA 30975 NOK
634-1088 634-1228
VWR® PV4 Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
The PV4 visible spectrophotometer is an easy to use instrument with advanced performance.
Highly accurate and reliable, with several basic functions, such as photometry test, quantitation test and spectrum scanning.
Leveringsinformasjon: Delivered with spectrophotometer, glass cuvettes (4 pcs.), power cord (Euro/UK/CH plug), software USB drive (1 pc.), USB cable (1 pc.), quick manual (1 pc.), dust cover (1 pc.), fuse (1 pc.).