VWR® P4, UV/Visible Spectrophotometer
Leverandør: VWR Collection
634-1229EA 56900 NOK
634-1229
VWR® P4, UV/Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
The P4 UV/Visible spectrophotometer is an easy to use instrument with advanced performance.
  • Highly accurate and reliable
  • Simple, elegant design with aluminum base
  • Easy to change lamp, user friendly interface
Provides several basic functions, such as photometry test, quantitation test and spectrum scanning.
Leveringsinformasjon: Delivered with spectrophotometer, glass cuvettes (4 pcs), power cord (Euro/UK/CH plug), software USB drive (1PC), USB cable (1PC), quick manual (1PC), dust cover (1PC), fuse (1PC).