VWR® P6, UV/Visible Spectrophotometer
Leverandør: VWR Collection
634-1230EA 38451 NOK
634-1230
VWR® P6, UV/Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
The P6 UV/Visible single beam spectrophotometer is an easy to use instrument with advanced performance.
  • Highly accurate and reliable
  • Simple, elegant design with aluminum base
  • Wireless tablet control
  • Easy to change lamp
  • User friendly interface
  • 64 GB internal storage capabilites
  • USB-C, WI-FI, Bluetooth, HDMI connectivity
Provides several basic functions, such as photometry test, multi wavelength scanning, kinetics, quantitation test, spectrum scanning and biological methods.
The instrument is shipped with VWR® UV studio standard software, sampleMore Product Information
Limited Time Sale
Select items below are currently on sale
Bestill nå
SPESIFIKASJONER
Modell P6
Environmentally Preferable
Absorberingsområde –0,3 - +3,0 A
Photometric range 0 - 200% T
Wavelength accuracy ±0,5 nm
Bølgelengdereproduserbarhet ≤0,3 nm
Lyskilde Tungsten/deuterium
Skannehastighet 5000 nm/min
Spektral båndbredde 1,5 nm
B×D×H 456×360×185 mm
Vekt 11 kg