VWR® V-1200 Visible Spectrophotometer
Leverandør: VWR Collection
634-6000EA 24225 NOK
634-6000
VWR® V-1200 Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
The V-1200 is designed for routine, daily, quality control requirements, in-process control and teaching laboratories.
  • Large LCD (128×64 P)
  • Can save up to a total of 200 results and 200 standard curves
  • Download data to PC via USB interface
With the support of the standard software, stored results and data can be downloaded from internal memory to PC.
Leveringsinformasjon: Supplied with four optical glass cells, four-position cell changer, power cable with UK and EU plug and dust cover.