VWR® UV-1600PC, UV/Visible Spectrophotometer
Leverandør: VWR Collection
634-6001EA 39450 NOK
634-6001
VWR® UV-1600PC, UV/Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
The UV-1600PC is designed for routine daily quality control requirements, in-process control and teaching laboratories.
  • Can save up to a total of 200 results and 200 standard curves
  • Download data to PC via USB interface
  • Optional accessories including an automatic eight-cell changer
Programvaren tilbyr de viktigste metodene for rutinemålinger.
Leveringsinformasjon: Supplied with four optical glass cells, two quartz cells, four-position cell holder, power cable with UK and EU plug, dust cover and application software for PC control.
Bestill nå
SPESIFIKASJONER
Modell UV-1600PC
Environmentally Preferable
Målemoduser Photometry, quantitation, kinetics
Display Dot matrix LCD (128×64 P)
Photometric accuracy ±0,5%T or 0,005A @ 1A
Photometric range 0 - 200% T, -0,3 - 3,0 A, 0 - 9999 C
Fotometrisk stabilitet ±0,002 A/h (500 nm)
Wavelength accuracy ±0,5 nm
Wavelength range 190 - 1100 nm
Bølgelengdeområde 190 − 1100 nm
Bølgelengdereproduserbarhet ≤0,3 nm
Detektor Grating 1200 lines/mm silicon photodiode
Elektronisk AC 110/220 V, 50/60 Hz
Grensesnitt USB, parallel
Språk English, German, Spanish, French
Lyskilde Tungsten
Minne 200 results and 200 standard curves
Utbytte USB
Spektral båndbredde 4 nm
Strølys ±0,05% T (220; 360 nm)
B×D×H 490×360×240 mm
Vekt 14 kg