VWR® UV-6300PC, UV/Visible Spectrophotometer
Leverandør: VWR Collection
634-6041EA 75925 NOK
634-6041
VWR® UV-6300PC, UV/Visible Spectrophotometer
Spektrofotometre og fotometre UV/Vis-spektrofotometere
UV/Visible spectrophotometer with double-beam optical system, designed for applications including general research, pharmaceutical, biochemical and clinical laboratory use, new material development and quality control.
  • Standard photometrics: Absorption, transmission and concentration
  • Quantitative analysis and kinetics, multi-wavelength scan and spectrum scan
  • DNA/RNA and protein analysis
  • Internal control via keypad and PC control (optional)
  • UV-Vis Analyst software for more powerful data processing and expanded data collecting
Can be used in stand-aloneMore Product Information