VWR® UV-3100PC, Spektrofotometer UV/Vis
Leverandør: VWR Collection
634-6042EA 58725 NOK
634-6042
VWR® UV-3100PC, Spektrofotometer UV/Vis
Spektrofotometre og fotometre UV/Vis-spektrofotometere
UV/Visible spectrophotometer for advanced measurements in quality control, biochemical and pharmaceutical applications.
  • Absorption, transmission and concentration photometry
  • Up to 10 quantitative standards
  • Four methods for calibration curve: Linear - linear through zero - square - cubic
  • Off-line calculation, re-scaling, curve tracking and peak search
  • Kinetic scanning time course or reacting rate calculations
Leveringsinformasjon: Leveres med 4 celler av optisk glass, 2 quartzceller, støvdeksel, USB-kabel, programvare for PC-kontroll, cellehoder med 4More Product Information