Rack for cuvettes
Leverandør: Brand
759500
634-8100EA 265 NOK
634-8100
Rack for cuvettes
Stativer for rør Kyvettestativer
PP.
  • Numbered positions to aid sample identification
  • Autoclavable at 121 °C
Rack with numbered positions for 2×8 cuvettes of 10 mm path length.