Macro cuvette
Leverandør: Greiner Bio-One
614101_1000
634-8211EA 1950 NOK
634-8211
Macro cuvette
Kyvetter Spektrofotometerkyvetter
Designed for enzymatic determinations, since the very thin walls enable rapid and uniform tempering.
  • Manufactured from crystal clear polystyrene
  • Characterised by minimal light scatter
  • High transmission