Absorption Cells with Lids
Leverandør: HELLMA
114-10-40 104-50-40 104-10-46 104-10-40 108-000-10-40 104-10-K-40 104-50-K-40
634-9158EA 4850 NOK
634-9158 634-9125 634-9132 634-9122 634-9154 634-0849 HELL104-50-K-40
Absorption Cells with Lids
Kyvetter Spektrofotometerkyvetter
Hellma® high precision quartz glass semi-micro cells for absorption measurements in the UV/Vis range, 10 to 50 mm path length, with lid.
  • Cells with PTFE lid
  • 10 to 50 mm path length
  • Highest precision
  • Excellent transmission
  • Sustainably produced
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.