Refractometer test plates and contact liquids
Leverandør: BELLINGHAM STANLEY
10-43
635-0000EA 1100 NOK
635-0000
Refractometer test plates and contact liquids
Standarder Brytningsindeks standarder
Solid test plates offer a convenient method of calibration and verifying the performance of optical handheld and Abbe refractometers.
  • Silica test plates for added traceability
  • Glass test plates for honey handheld refractometers
  • No temperature correction required
  • Inert material offering longevity
Depending on the type of instrument and the scale range to be verified, a choice can be made between glass or silica test plates. Silica test plates are traceable to recognised standards and may be recertified as part of a quality control standard operating procedure.
Leveringsinformasjon: Supplied with a certificate of verification. Additional certification for silica plates is available as an optional extra.