Printer, impact, CBM910
Leverandør: BELLINGHAM STANLEY
55-14
635-0478EA 5575 NOK
635-0478
Printer, impact, CBM910
Printere Document Printers
Dot matrix, miniature, 24 column, RS232, impact printer that can be used to provide hard copy of refractometer or polarimeter measurement data for FDA regulated environments or GLP audit trail as dot matrix impact printers are the preferred option for data longevity compared to thermal type printers.
  • Dot matrix for longevity of print
  • 24 column width
  • Small footprint
Bellingham + Stanley RFM refractometers and ADP polarimeters print outputs are optimised to print with this 24-column printer, making this a convenient printer for all models.