Pocket refractometer, DEF-Chek
Leverandør: REICHERT
13940013
635-0507EA 7725 NOK
635-0507
Pocket refractometer, DEF-Chek
Refraktometere Refraktometere, håndholdte
Selective catalytic reduction systems for diesel trucks and buses requires Diesel Exhaust Fluid (DEF) at an exact concentration of 32,5 % urea.
The DEF-Chek offers quick accurate results.