Portable refractometer for automotive care, Multi-Chek
Leverandør: REICHERT
13940015
635-0509EA 7725 NOK
635-0509
Portable refractometer for automotive care, Multi-Chek
Refraktometere Refraktometere, håndholdte
Three-in-one tester for measuring freezing point of engine coolants with ATC (ethylene glycol and propylene glycol); the specific gravity and condition of battery electrolytes and lastly the freezing point of the wind screen washer fluid.