Pocket digital refractometer, Brake-Chek
Leverandør: REICHERT
13940017
635-0510EA 7725 NOK
635-0510
Pocket digital refractometer, Brake-Chek
Refraktometere Refraktometere, håndholdte
To measure the key fluids for automotive to help maintain safe operation on the road. Test brake fluids by measuring the boiling point to indicate the extent of the breakdown of the fluid or increase in water content.
  • Distilled water calibration
  • Glass prism
Sertifiseringer: IP 65, RoHS and WEEE
Leveringsinformasjon: Supplied with two AAA batteries for 10 000 readings.