Pocket acidity meter, PAL-BX
Leverandør: Atago
7102
635-0729EA 7825 NOK
635-0729
Pocket acidity meter, PAL-BX
Refraktometere Refraktometere, håndholdte
The PAL-BX pocket acidity meter is specifically for measuring citrus in grapes and wine.
  • Features with backlit screen
  • Accurate and easy measurements
  • Measure Brix and acidity within one unit
Sertifiseringer: IP 65
Bestillingsinformasjon: Includes a digital scale, plastic pipettes, one beaker and one measuring spoon.