Handheld refractometer, Acidity meter for vinegar, PAL- Easy ACID 181 master kit
Leverandør: Atago
7781
635-0833EA 5250 NOK
635-0833
Handheld refractometer, Acidity meter for vinegar, PAL- Easy ACID 181 master kit
Refraktometere Refraktometere, håndholdte
This Master Kit is specifically for measuring vinegar and includes everything you need for easy, precise and accurate measurements.
  • ATAGO's durable titan electrode will show the result that correlated with that of titaration
To measure acidity, sample needs to be dilulted.
Bestillingsinformasjon: Requires 2 x AAA alkaline batteries.
Leveringsinformasjon: The master kit comes complete with a scale, 1 beaker, and 1 measuring spoon.