Handheld refractometer, OE 200
Leverandør: REICHERT
137528L0
635-2115EA 4225 NOK
635-2115
Handheld refractometer, OE 200
Refraktometere Refraktometere, håndholdte
This handheld refractometer for wine provides fast and accurate measurements of dissolved sugars when testing grapes, citrus fruits, fruit juices, soft drinks, melons and maple sap for the winemaking industry.