Pocket refractometer, Brix/RI-Chek
Leverandør: REICHERT
13940000
635-2530EA 6175 NOK
635-2530
Pocket refractometer, Brix/RI-Chek
Refraktometere Refraktometere, håndholdte
The Brix/RI-Chek is a battery operated pocket refractometer.
  • Automatic temperature compensation (ATC)
  • Simple single-handed operation
  • Delivers rapid results
Sertifiseringer: IP 65 allows rinsing off the sample with running water.