Handheld refractometer, Rhino VET 360
Leverandør: REICHERT
137536L0
635-2542EA 3650 NOK
635-2542
Handheld refractometer, Rhino VET 360
Refraktometere Refraktometere, håndholdte
Shock resistant refractometer for urine specific gravity and protein concentration.
  • Test takes seconds, performs without battery
  • Automatic temperature compensation (ATC) for accuracy
  • Water resistant, dustproof and sealed internal optics
The Rhino VET 360 is used for plasma, serum and peritoneal fluids of veterinary clinical applications.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Nøyaktighet Urine ±0,001 SG ; Serum ±0,2 g/100 ml
B×D×H 235×38×38 mm
Vekt 162 g