Formaldehyd 4% in aqueous solution, VOLUSOL®, buffered pH 7.2 -7.6

Leverandør: VWR Chemicals
Danger


Stabilisering: stabilisert med metanol 0,5-1,5 %

6368.5000EA 1575 NOK
6368.5000
Formaldehyd 4% in aqueous solution, VOLUSOL®, buffered pH 7.2 -7.6
Formaldehyd
Formel: CH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8

Bestill nå

Spesifikasjon Testresultater

Assay (Formaldehyde) 3.5 --> 4.5 %
pH (20°C) 7.2 --> 7.6
Methanol 0.5 --> 1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR