Bloom test vessel
Leverandør: DWK Life Sciences
211250105
646-0035EA 1375 NOK
646-0035
Bloom test vessel
Reaksjonskolber Reaksjonskolber, begerglassformet
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Manufactured according to DIN ISO 9665.