Trietylamin ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™

Leverandør: Honeywell Chemicals
Danger

65897-50ML
65897-50MLEA 1075 NOK
65897-50ML
Trietylamin ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
Trietylamin

Eluent additive for LC-MS.

Formel: (C₂H₅)₃N
Kokepunkt: 90 °C
Smeltepunkt: –115 °C
Tetthet: 0,73 g/cm3 (20 °C)
Flash punkt: –11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009051
CAS-nummer: 121-44-8
EINECS: 231-913-4
FN: 1296
ADR: 3,II
Merck Indeks: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Identity (IR) complying
Assay (GC) Min. 99.5 %
Refractive index (n 20/D) 1.400 - 1.402
Appearance complying
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.5 %
Aluminium (Al) Max. 0.2 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
UV- cutoff 250.0 - 280.0 nm
Vapor Density 3.5 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR