Titrators, Karl Fischer, coulometric, TitroLine® 7500 KF Trace
Leverandør: SI Analytics
285220860 285220870 285220880 285220890 285227800
661-0141EA 0 NOK
661-0141 661-0142 661-0143 661-0144 661-0290
Titrators, Karl Fischer, coulometric, TitroLine® 7500 KF Trace
Titratorer
These very compact, easy to operate instruments are ideal for coulometric water determination according to Karl Fischer in low water content, as well as for bromine number determination. They are suitable for small liquid or gaseous sample amounts, and can be used for quality control in the chemical, petrochemical, pharmaceutical and food industries.
  • High visibility, full colour display with online titration curve
  • Standard methods for different applications (for example blank value)
  • Include three USB and two RS232 ports for expansion and connection of