pH indicator papers, universal/special
Leverandør: MACHEREY-NAGEL
90201 90202 90203 90205 90206 90207 90208 90209 90210 90211 90212 90213 90225 90226 90227 90228 90229 90230 90231 90233 90224 90204 90214 90232 90234
662-0209EA 325 NOK
662-0209 662-0230 662-1866 662-0231 MANA90206 662-0206 MANA90208 662-0232 662-0233 MANA90211 MANA90212 MANA90213 MANA90225 MANA90226 MANA90227 MANA90228 MANA90229 MANA90230 MANA90231 MANA90233 662-0193 662-0204 MANA90214 MANA90232 MANA90234
pH indicator papers, universal/special
Testpapir pH-indikatorstrips
pH indicator papers have been available for decades and are the standard for many applications. For each pH value these papers show a single colour which can be matched with the colour scale at intervals of 0,2 to 1 pH units.
  • Long term stable and protected against external influences due to plastic reel
  • No accessories needed - always ready to use
  • Reliable results due to easy and accurate read-off from colour scale
  • Indicator papers are supplied as rolls with 5 m length and 7 mm width
pH indicators are manufactured from high quality filter papers. TheMore Product Information