pH electrode, micro, Orion™ 9863BN
Leverandør: Thermo Orion
9863BN
662-0889EA 18350 NOK
662-0889
pH electrode, micro, Orion™ 9863BN
Elektroder pH-elektroder
This bevel needle-tipped 16-gauge glass body combination electrode can be used to measure pH by piercing septa.
  • Made of very thin, delicate glass
  • Single junction Ag/AgCl
  • Refillable