VWR® Traceable®, Light Meter, Dual-Display
Leverandør: VWR Collection
662-0940EA 4800 NOK
662-0940
VWR® Traceable®, Light Meter, Dual-Display
Lysmålere
Dual-displays show lux or foot-candles, or percent differential from a reference point, and the selected light source (daylight, tungsten, fluorescent or mercury). Traceable® light meter can be connected to a computer or data logger for monitoring and keeping results. To ensure accuracy, an individually serial numbered Traceable® certificate is provided from ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
  • Two displays show lux or foot-candles
  • Computer output
  • Data hold button '