VWR® pHenomenal®, pH-elektroder
Leverandør: VWR Collection
620-1659EA 2300 NOK
620-1659 662-1162 662-1163 662-1164 662-1158 662-1159 662-1161 662-1156 662-1157 662-1247
VWR® pHenomenal®, pH-elektroder
Elektroder pH-elektroder
pH-elektroder med BNC-støpsel og -kabel.
These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.
* Integrated temperature probe.
Leveringsinformasjon: Includes pH-electrode only. 620-1659 includes pHenomenal® TEMP21 T-sensor (NTC 30 kOhm) only.