pH-elektrode, Sentix® IDS
Leverandør: WTW
103740 103780 103750 103741
662-1249EA 5500 NOK
662-1249 662-1254 662-1253 662-1250
pH-elektrode, Sentix® IDS
Elektroder pH-elektroder
Utvidet sensorkvalitet kombinert med den mest moderne måleelektronikken.
  • Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • Measurement integrity benchmarking of the electrodes by QSC (Quality Sensor Control)
  • IDS electrodes store their serial number and their calibration records
Ø×L: 12×120 mm
Bestillingsinformasjon: Ulike kabellengder er tilgjengelig